Thermo Scientific Orion VERSA STAR Advanced Meter
Modularity allows you to configure the meter how you need it--for now and for later
  • Customize the four measurement channels with five different parameter modules
  • Large color graphic LCD with intuitive multilanguage interface
  • Non-volatle memory holds up to 2000 data points with time and date stamp
  • Easy-to-operate USB and RS232 ports and data transfer software included
The VERSA STAR advanced meter provides the versatility to operate from one to four measurement channels at the same time. The five parameter modules available for the channels including; pH, pH/ISE, Conductivity, DO/RDO, and pH with LogR. Customize the meter you need with the options of meter only and modules, a meter with module package, or a full kit with meter, modules, electrode and solutions.
This versatile meter has a large color graphic LCD display. The informative display shows readings with temperature, connections, time, date, sample ID, user ID and calibration data. Plain language screen prompts, soft keys, and a multilanguage interface supports English, Spanish, French, Italian, German, and Chinese.
Non-volatle memory holds up to 2000 data sets with time and date stamp. Easy-to-operate USB and RS232 ports and data transfer software take stress out of data management.
341,710.00 - ₹927,000.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Waterproof Description
Availability Pricing
Thermo Scientific VSTAR10 Versa Star with pH Module 5882581 Mfr # VSTAR10
COMPARE
Yes Versa Star with pH Module
₹544,000.00INR / Each
Thermo Scientific VSTAR22 Versa Star with Conductivity Module Kit 5882584 Mfr # VSTAR22
COMPARE
Yes Versa Star with Conductivity Module Kit
₹864,000.00INR / Each
Thermo Scientific VSTAR40A2 Versa Star with pH/ISE Module Kit 5882588 Mfr # VSTAR40A2
COMPARE
Yes Versa Star with pH/ISE Module Kit
₹927,000.00INR / Each
Thermo Scientific VSTAR40B Versa Star with Two pH/ISE Modules 5882589 Mfr # VSTAR40B
COMPARE
Yes Versa Star with Two pH/ISE Modules
₹873,000.00INR / Each
Thermo Scientific VSTAR50 Versa Star with pH/Conductivity Modules 5882591 Mfr # VSTAR50
COMPARE
Yes Versa Star with pH/Conductivity Modules
₹372,290.00INR / Each
Thermo Scientific VSTAR52 Versa Star with pH/Conductivity Modules Kit 5882592 Mfr # VSTAR52
COMPARE
Yes Versa Star with pH/Conductivity Modules Kit
₹444,720.00INR / Each
Thermo Scientific VSTAR80 Versa Star withpH/LogR Modules 5882593 Mfr # VSTAR80
COMPARE
Yes Versa Star withpH/LogR Modules
₹341,710.00INR / Each
Thermo Scientific VSTAR82 Versa Star withpH/LogR Modules Kit 5882594 Mfr # VSTAR82
COMPARE
Yes Versa Star withpH/LogR Modules Kit
₹411,940.00INR / Each
Thermo Scientific VSTAR90 Versa Star with pH/ISE, Cond, RDO/DO Modules 5882595 Mfr # VSTAR90
COMPARE
Yes Versa Star with pH/ISE, Cond, RDO/DO Modules
₹562,220.00INR / Each
Thermo Scientific VSTAR92 Versa Star with pH/ISE, Cond, RDO/DO Modules Kit 5882596 Mfr # VSTAR92
COMPARE
Yes Versa Star with pH/ISE, Cond, RDO/DO Modules Kit
Call For Price
Showing 1- 10 of 10