VERSA STAR meters include: an electrode arm with redesigned holder, and a universal power adapter.
Key Features
Customize the four measurement channels with five different parameter modules
Large color graphic LCD with intuitive multilanguage interface
Non-volatle memory holds up to 2000 data points with time and date stamp
Easy-to-operate USB and RS232 ports and data transfer software included
Up to 5 point calibration with automatic recognition for USA/NIST and DIN buffers
Editing calibration errors with a one point recalibration
Selectable cell constant for 2- or 4-cell conductivity probes
Selectable reading reference temperature of 5, 10, 15, 20 or 25°C
Choice of practical salinity or natural sea water curves for salinity
Accepts both polarographic and RDO(optical/luminessence) probes
Automatic temperature and barometic pressure compensation, manual salinity compensation
3 year warranty
More About this Item
The VERSA STAR advanced meter provides the versatility to operate from one to four measurement channels at the same time. The five parameter modules available for the channels including; pH, pH/ISE, Conductivity, DO/RDO, and pH with LogR. Customize the meter you need with the options of meter only and modules, a meter with module package, or a full kit with meter, modules, electrode and solutions.
This versatile meter has a large color graphic LCD display. The informative display shows readings with temperature, connections, time, date, sample ID, user ID and calibration data. Plain language screen prompts, soft keys, and a multilanguage interface supports English, Spanish, French, Italian, German, and Chinese.
Non-volatle memory holds up to 2000 data sets with time and date stamp. Easy-to-operate USB and RS232 ports and data transfer software take stress out of data management.
Adds one pH/ISE, one Conductivity, and one DO/RDO measurement modules. Probes sold separately.
Specifications
Meter TypeBenchtop
Display typeGraphic LCD
Min pH (pH)-2
Max pH (pH)20
pH Resolution0.001/0.01/0.1
pH Accuracy±0.002 pH
Buffer RecognitionDIN, NIST, and USA
CalibrationUp to 6 points
Min mV (mV)-2000
Max mV (mV)2000
mV Resolution0.1 or 1 mV
mV Accuracy±0.2 mV or 0.05%, whichever is greater
Min ISE0 to 19999 ppm
ISE Resolution0.01 ppm
ISE Accuracy±0.05% of reading
Conductivity Accuracy±0.5% FS +1 LSD
Min TDS0 to 200 ppt
Min Salinity0 to 80 ppt
Salinity Accuracy±0.1 ppt
Resistivity Accuracy±0.5% ±1 digit
Min Dissolved Oxygen0 to 90 ppm; 0 to 600 %
Dissolved Oxygen Accuracy±0.2 ppm
Min Temperature (° C)-5
Max Temperature (° C)105
Min Temperature (° F)23
Max Temperature (° F)221
InterfaceRS-232 and USB
WaterproofYes
Power (VAC)90 to 260
Power (Hz)50 or 60
Battery4 AA (not included)
Width (cm)17.78
Height (cm)24.765
Width (in)7
Length (in)3 3/8
Height (in)9 3/4
DescriptionVersa Star with pH/ISE, Cond, RDO/DO Modules