Thermo Scientific VSTAR82 Versa Star withpH/LogR Modules Kit
Representative image only.
Thermo Scientific VSTAR82 Versa Star withpH/LogR Modules Kit
Thermo Scientific | Mfr: VSTAR82| Item: 5882594
Modularity allows you to configure the meter how you need it--for now and for later
₹411,940.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹411,940.00INR / Each
Buy It Together
Current Item
Thermo Scientific VSTAR82 Versa Star withpH/LogR Modules Kit
Thermo Scientific VSTAR82 Versa Star withpH/LogR Modules Kit
5882594
₹411,940.00INR / Each
+ =
Total Price 436640.00 ADD 2 ITEMS TO CART
Product Description
Includes
VERSA STAR meters include: an electrode arm with redesigned holder, and a universal power adapter.
Key Features
  • Customize the four measurement channels with five different parameter modules
  • Large color graphic LCD with intuitive multilanguage interface
  • Non-volatle memory holds up to 2000 data points with time and date stamp
  • Easy-to-operate USB and RS232 ports and data transfer software included
  • Up to 5 point calibration with automatic recognition for USA/NIST and DIN buffers
  • Editing calibration errors with a one point recalibration
  • The most advanced electrode diagnostics
  • No temperature
  • 3 year warranty
More About this Item
The VERSA STAR advanced meter provides the versatility to operate from one to four measurement channels at the same time. The five parameter modules available for the channels including; pH, pH/ISE, Conductivity, DO/RDO, and pH with LogR. Customize the meter you need with the options of meter only and modules, a meter with module package, or a full kit with meter, modules, electrode and solutions.
This versatile meter has a large color graphic LCD display. The informative display shows readings with temperature, connections, time, date, sample ID, user ID and calibration data. Plain language screen prompts, soft keys, and a multilanguage interface supports English, Spanish, French, Italian, German, and Chinese.
Non-volatle memory holds up to 2000 data sets with time and date stamp. Easy-to-operate USB and RS232 ports and data transfer software take stress out of data management.
Adds one pH logR module, ROSS SureFlow pH electrode (58824-40), ATC probe (58824-48), pH buffer kit with 475 mL each of pH 4, 7, and 10 buffer.
3 Year WarrantyIP54 Rated
Specifications
  • Meter TypeBenchtop
  • WaterproofYes
  • DescriptionVersa Star withpH/LogR Modules Kit