Thermo Scientific VSTAR92 Versa Star with pH/ISE, Cond, RDO/DO Modules Kit

Representative image only.

Thermo Scientific VSTAR92 Versa Star with pH/ISE, Cond, RDO/DO Modules Kit

Thermo Scientific | Mfr: VSTAR92| Item: 5882596

Modularity allows you to configure the meter how you need it--for now and for later

Call For Price

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

Call For Price

Buy It Together

Current Item

Thermo Scientific VSTAR92 Versa Star with pH/ISE, Cond, RDO/DO Modules Kit
Thermo Scientific VSTAR92 Versa Star with pH/ISE, Cond, RDO/DO Modules Kit
5882596

Call For Price
+ =
Total Price 24700.00 ADD 2 ITEMS TO CART
Product Description

Includes

VERSA STAR meters include: an electrode arm with redesigned holder, and a universal power adapter.

Key Features

  • Customize the four measurement channels with five different parameter modules
  • Large color graphic LCD with intuitive multilanguage interface
  • Non-volatle memory holds up to 2000 data points with time and date stamp
  • Easy-to-operate USB and RS232 ports and data transfer software included
  • Up to 5 point calibration with automatic recognition for USA/NIST and DIN buffers
  • Editing calibration errors with a one point recalibration
  • Selectable cell constant for 2- or 4-cell conductivity probes
  • Selectable reading reference temperature of 5, 10, 15, 20 or 25°C
  • Choice of practical salinity or natural sea water curves for salinity
  • Accepts both polarographic and RDO(optical/luminessence) probes
  • Automatic temperature and barometic pressure compensation, manual salinity compensation
  • 3 year warranty

More About this Item

The VERSA STAR advanced meter provides the versatility to operate from one to four measurement channels at the same time. The five parameter modules available for the channels including; pH, pH/ISE, Conductivity, DO/RDO, and pH with LogR. Customize the meter you need with the options of meter only and modules, a meter with module package, or a full kit with meter, modules, electrode and solutions.

This versatile meter has a large color graphic LCD display. The informative display shows readings with temperature, connections, time, date, sample ID, user ID and calibration data. Plain language screen prompts, soft keys, and a multilanguage interface supports English, Spanish, French, Italian, German, and Chinese.

Non-volatle memory holds up to 2000 data sets with time and date stamp. Easy-to-operate USB and RS232 ports and data transfer software take stress out of data management.

Adds one pH/ISE, one Conductivity, and one DO/RDO measurement modules. ROSS Triode pH/ATC electrode (05718-34); Buffer kit with 475 mL each of 4, 7, and 10 pH buffer (53350-66);DuraProbe conductivity cell (19704-70); and five 60-mL bottles of 1413 µS standard; Orion polarographic DO probe (53101-60), DO probe maintenance kit (53107-55), BOD adapters

3 Year WarrantyIP54 Rated
Specifications
  • Meter TypeBenchtop
  • Display typeGraphic LCD
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • Buffer RecognitionDIN, NIST, and USA
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 or 1 mV
  • Min ISE0 to 19999 ppm
  • ISE Resolution0.01 ppm
  • ISE Accuracy±0.05% of reading
  • Min TDS0 to 200 ppt
  • Min Salinity0 to 80 ppt
  • Salinity Accuracy±0.1 ppt
  • Resistivity Accuracy±0.5% ±1 digit
  • Min Dissolved Oxygen0 to 90 ppm; 0 to 600 %
  • Dissolved Oxygen Accuracy±0.2 ppm
  • Min Temperature (° C)-5
  • Max Temperature (° C)105
  • Min Temperature (° F)23
  • Max Temperature (° F)221
  • InterfaceRS-232 and USB
  • WaterproofYes
  • Power (VAC)90 to 260
  • Power (Hz)50 or 60
  • Battery4 AA (not included)
  • DescriptionVersa Star with pH/ISE, Cond, RDO/DO Modules Kit
  • Warranty3 year
Reviews
Q&A