Thermo Scientific Nalgene Rectangular Carboys with Handle, PC
Don't lose sight of your solution
  • Transparent PC allows easy viewing of solutions
  • Easy trasport via steel handle
  • Autoclavable
Space-saving, leakproof polycarbonate (PC) carboys feature liter/gallon graduations and built-in shoulder loops with steel handles. Models with quick-action spigots accept 5⁄8" ID tubing and come with adapters for 1⁄4" to 1⁄2" ID tubing, integrally molded threaded boss, and two PTFE O-rings for additional versatility.
82,300.00 - ₹106,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Cap / Stopper Size Material
Availability Pricing
Thermo Scientific Nalgene DS2213-0050 rectangular polycarbonate carboy with handle, 20 L 0608470 Mfr # DS2213-0050
COMPARE
20 100 mm Polycarbonate(PC)
₹106,000.00INR / Each
Thermo Scientific Nalgene 2322-0020 Rectangular Carboy with Spigot PC, 9 L, 6/Cs 0608480 Mfr # 2322-0020
COMPARE
9 100 mm Polycarbonate(PC)
₹82,300.00INR / Each
Thermo Scientific Nalgene 2322-0050 Rectangular Carboy with Spigot PC, 20 L, 4/Cs 0608490 Mfr # 2322-0050
COMPARE
20 100 mm Polycarbonate(PC)
₹104,000.00INR / Each