Slide Preparation System
Mix-and-match components for flexible slide staining
Staining Dishes and Covers accommodate slides measuring 75 x 25 mm (3" x 1"), 75 x 38 mm (3" x 11⁄2"), and 75 x 51 mm (3" x 2"). Approximate inside dimensions are 118 mm L x 88 mm W x 70 mm H. Order racks separately.
Rack 48585-76 holds 30 microscope slides. Handle is permanently attached, but hinged to permit closure of the dish and easy insertion and removal of microscope slides. Constructed of nontarnishing stainless steel (SS); resists most staining solutions.
Rack 48585-78 holds 16 single slides, 31 slides arranged alternately straight across and diagonally, or 32 slides back-to-back. Manufactured from soda-lime glass.
Rack 48585-80 holds 20 single slides, 39 slides arranged alternately straight across and diagonally, or 40 slides back-to-back. Manufactured from soda-lime glass.
16,800.00 - ₹32,900.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
DWK Life Sciences (Wheaton) 900303 Slide Preparation System Glass Staining Dish and Cover 4858570 Mfr # 900303
COMPARE
Slide Preparation System Glass Staining Dish and Cover
₹32,900.00INR / Each
DWK Life Sciences (Wheaton) 900301 Slide Preparation Dish for staining system 48585-70 4858572 Mfr # 900301
COMPARE
Slide Preparation Dish for staining system 48585-70
₹24,700.00INR / Each
DWK Life Sciences (Wheaton) 900234 Slide Preparation System Stainless Steel Staining Rack for 30-Slides 4858576 Mfr # 900234
COMPARE
Slide Preparation System Stainless Steel Staining Rack for 30-Slides
₹18,400.00INR / Each
DWK Life Sciences (Wheaton) 900254 Glass Staining Rack Slide Preparation System for 16 to 32 Slides 4858578 Mfr # 900254
COMPARE
Glass Staining Rack Slide Preparation System for 16 to 32 Slides
₹16,800.00INR / Each
DWK Life Sciences (Wheaton) 900304 Glass Staining Rack Slide Preparation System for 20 to 40 Slides 4858580 Mfr # 900304
COMPARE
Glass Staining Rack Slide Preparation System for 20 to 40 Slides
₹23,000.00INR / Each