DWK Life Sciences (Wheaton) 900254 Glass Staining Rack Slide Preparation System for 16 to 32 Slides
Representative image only.
DWK Life Sciences (Wheaton) 900254 Glass Staining Rack Slide Preparation System for 16 to 32 Slides
DWK Life Sciences (Wheaton) | Mfr: 900254| Item: 4858578
Mix-and-match components for flexible slide staining
₹16,800.00INR / Each
₹16,800.00INR / Each
Buy It Together
Current Item
DWK Life Sciences (Wheaton) 900254 Glass Staining Rack Slide Preparation System for 16 to 32 Slides
DWK Life Sciences (Wheaton) 900254 Glass Staining Rack Slide Preparation System for 16 to 32 Slides
4858578
₹16,800.00INR / Each
+ + =
Total Price 166200.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Staining Dishes and Covers accommodate slides measuring 75 x 25 mm (3" x 1"), 75 x 38 mm (3" x 11⁄2"), and 75 x 51 mm (3" x 2"). Approximate inside dimensions are 118 mm L x 88 mm W x 70 mm H. Order racks separately.
Rack 48585-76 holds 30 microscope slides. Handle is permanently attached, but hinged to permit closure of the dish and easy insertion and removal of microscope slides. Constructed of nontarnishing stainless steel (SS); resists most staining solutions.
Rack 48585-78 holds 16 single slides, 31 slides arranged alternately straight across and diagonally, or 32 slides back-to-back. Manufactured from soda-lime glass.
Rack 48585-80 holds 20 single slides, 39 slides arranged alternately straight across and diagonally, or 40 slides back-to-back. Manufactured from soda-lime glass.
Specifications
  • Capacity (Number of slides)32
  • DescriptionGlass Staining Rack Slide Preparation System for 16 to 32 Slides