DWK Life Sciences (Wheaton) Staining Dish System with Glass Rack and Cover; 20-Slide Capacity
Representative image only.
DWK Life Sciences (Wheaton) Staining Dish System with Glass Rack and Cover; 20-Slide Capacity
DWK Life Sciences (Wheaton) | Mfr: 900200| Item: 4858500
Designed for secure staining of microscope slides
₹28,400.00INR / Each
₹28,400.00INR / Each
Buy It Together
Current Item
DWK Life Sciences (Wheaton) Staining Dish System with Glass Rack and Cover; 20-Slide Capacity
DWK Life Sciences (Wheaton) Staining Dish System with Glass Rack and Cover; 20-Slide Capacity
4858500
₹28,400.00INR / Each
+ + + =
Total Price 126800.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Rack handle allows for convenient moving around the lab
  • Complete with dish, cover, and glass rack
More About this Item
Choose the staining dish and rack system that meets your application requirements. Accommodate slides measuring 75 x 25 mm (3" x 1"), 75 x 38 mm (3" x 1 1⁄2"), and 75 x 51 mm (3" x 2").
This 20-slide unit is the standard for manual staining procedures. The removable glass slide rack has an open bottom to facilitate rapid immersion and drainage, reducing carryover. The rack holds 10 single slides, 19 slides arranged alternately straight across and diagonally, or 20 slides back-to-back of standard size: 3" x 1" (75 x 25 mm), 3" x 1-1/2" (75 x 38 mm) and 3" x 2" (75 x 51 mm) sizes. Manufactured from soda-lime glass. Approximate inside dimensions: 95 mm L x 76 mm W x 64 mm D.
Specifications
  • Capacity (Number of slides)20
  • Internal Width (cm)7.1
  • Internal Height (cm)9.1
  • Internal Depth (cm)6
  • DescriptionStaining Dish System with Glass Rack and Cover; 20-Slide Capacity