WHEATON® Coplin Staining Jars with Cover
Ideal for staining slides
  • Constructed of durable soda-lime glass
  • Jar is designed to hold 3” x 1” (75mm x 25mm) slides
WHEATON® Coplin staining jars provide a stable container for staining slides. Glass covers protect slides from outside elements and reduce reagent evaporation. Compact size makes it easy to transfer slides and requires only a small volume of reagent.
10,300.00 - ₹11,200.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (Number of slides) Description
Availability Pricing
DWK Life Sciences (Wheaton) Coplin Staining Jar with Cover, Glass, 8-16 Slides 4858520 Mfr # 900620
COMPARE
8 single or 16 back-to-back Coplin Staining Jar with Cover, Glass, 8-16 Slides
₹10,300.00INR / Each
DWK Life Sciences (Wheaton) Coplin Staining Jar with Cover, Glass, 5-10 Slides 4858530 Mfr # 900470
COMPARE
5 single or 10 back-to-back Coplin Staining Jar with Cover, Glass, 5-10 Slides
₹11,200.00INR / Each