DWK Life Sciences (Wheaton) 900303 Slide Preparation System Glass Staining Dish and Cover
Representative image only.
DWK Life Sciences (Wheaton) 900303 Slide Preparation System Glass Staining Dish and Cover
DWK Life Sciences (Wheaton) | Mfr: 900303| Item: 4858570
Mix-and-match components for flexible slide staining
₹32,900.00INR / Each
₹32,900.00INR / Each
Buy It Together
Current Item
DWK Life Sciences (Wheaton) 900303 Slide Preparation System Glass Staining Dish and Cover
DWK Life Sciences (Wheaton) 900303 Slide Preparation System Glass Staining Dish and Cover
4858570
₹32,900.00INR / Each
+ + =
Total Price 182300.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Staining Dishes and Covers accommodate slides measuring 75 x 25 mm (3" x 1"), 75 x 38 mm (3" x 11⁄2"), and 75 x 51 mm (3" x 2"). Approximate inside dimensions are 118 mm L x 88 mm W x 70 mm H. Order racks separately.
Rack 48585-76 holds 30 microscope slides. Handle is permanently attached, but hinged to permit closure of the dish and easy insertion and removal of microscope slides. Constructed of nontarnishing stainless steel (SS); resists most staining solutions.
Rack 48585-78 holds 16 single slides, 31 slides arranged alternately straight across and diagonally, or 32 slides back-to-back. Manufactured from soda-lime glass.
Rack 48585-80 holds 20 single slides, 39 slides arranged alternately straight across and diagonally, or 40 slides back-to-back. Manufactured from soda-lime glass.
Specifications
  • DescriptionSlide Preparation System Glass Staining Dish and Cover