DWK Life Sciences (Wheaton) 900304 Glass Staining Rack Slide Preparation System for 20 to 40 Slides
Representative image only.
DWK Life Sciences (Wheaton) 900304 Glass Staining Rack Slide Preparation System for 20 to 40 Slides
DWK Life Sciences (Wheaton) | Mfr: 900304| Item: 4858580
Mix-and-match components for flexible slide staining
₹23,000.00INR / Each
₹23,000.00INR / Each
Buy It Together
Current Item
DWK Life Sciences (Wheaton) 900304 Glass Staining Rack Slide Preparation System for 20 to 40 Slides
DWK Life Sciences (Wheaton) 900304 Glass Staining Rack Slide Preparation System for 20 to 40 Slides
4858580
₹23,000.00INR / Each
+ + =
Total Price 172400.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Staining Dishes and Covers accommodate slides measuring 75 x 25 mm (3" x 1"), 75 x 38 mm (3" x 11⁄2"), and 75 x 51 mm (3" x 2"). Approximate inside dimensions are 118 mm L x 88 mm W x 70 mm H. Order racks separately.
Rack 48585-76 holds 30 microscope slides. Handle is permanently attached, but hinged to permit closure of the dish and easy insertion and removal of microscope slides. Constructed of nontarnishing stainless steel (SS); resists most staining solutions.
Rack 48585-78 holds 16 single slides, 31 slides arranged alternately straight across and diagonally, or 32 slides back-to-back. Manufactured from soda-lime glass.
Rack 48585-80 holds 20 single slides, 39 slides arranged alternately straight across and diagonally, or 40 slides back-to-back. Manufactured from soda-lime glass.
Specifications
  • Capacity (Number of slides)40
  • DescriptionGlass Staining Rack Slide Preparation System for 20 to 40 Slides