Scienceware Joint Thermometer adapters
For air-tight vacuum work. Threaded PP screw cap on top with an inside O-ring for fine depth adjustment of thermometers or tubing.
39,800.00 - ₹50,400.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Taper Size
Availability Pricing
Scienceware 20309-1680 Thermometer Adapter of PTFE, size 14/20 0614010 Mfr # 20309-1680
COMPARE
14/20
₹39,800.00INR / Each
Scienceware 20309-1710 Thermometer Adapter of PTFE, size 19/22 0614020 Mfr # 20309-1710
COMPARE
19/22
₹45,900.00INR / Each
Scienceware 20309-1716 Thermometer Adapter of PTFE, size 24/40 0614022 Mfr # 20309-1716
COMPARE
24/40
₹50,400.00INR / Each