Scienceware 20309-1680 Thermometer Adapter of PTFE, size 14/20
Representative image only.
Scienceware 20309-1680 Thermometer Adapter of PTFE, size 14/20
Scienceware | Mfr: 20309-1680| Item: 0614010
₹39,800.00INR / Each
₹39,800.00INR / Each
Product Description
More About this Item
For air-tight vacuum work. Threaded PP screw cap on top with an inside O-ring for fine depth adjustment of thermometers or tubing.
Specifications
  • Taper Size14/20
  • Port Size (ID) (mm)6 to 7
  • MaterialPolytetrafluoroethylene (PTFE)
  • DescriptionThermometer Adapter of PTFE, size 14/20