Scienceware 20309-1716 Thermometer Adapter of PTFE, size 24/40
Representative image only.
Scienceware 20309-1716 Thermometer Adapter of PTFE, size 24/40
Scienceware | Mfr: 20309-1716| Item: 0614022
₹50,400.00INR / Each
₹50,400.00INR / Each
Product Description
More About this Item
For air-tight vacuum work. Threaded PP screw cap on top with an inside O-ring for fine depth adjustment of thermometers or tubing.
Specifications
  • Taper Size24/40
  • Port Size (ID) (mm)6 to 7
  • MaterialPolytetrafluoroethylene (PTFE)
  • DescriptionThermometer Adapter of PTFE, size 24/40