Scienceware 20309-1710 Thermometer Adapter of PTFE, size 19/22
Representative image only.
Scienceware 20309-1710 Thermometer Adapter of PTFE, size 19/22
Scienceware | Mfr: 20309-1710| Item: 0614020
₹45,900.00INR / Each
₹45,900.00INR / Each
Product Description
More About this Item
For air-tight vacuum work. Threaded PP screw cap on top with an inside O-ring for fine depth adjustment of thermometers or tubing.
Specifications
  • Taper Size19/22
  • Port Size (ID) (mm)6 to 7
  • MaterialPolytetrafluoroethylene (PTFE)
  • DescriptionThermometer Adapter of PTFE, size 19/22