Sartorius MA-Series Infrared Moisture Analyzers
Ideal for quick and reliable determination of the moisture content of liquid, pasty, and solid substances
  • User-friendly programming operations with onscreen icons
  • Space-saving design conserves valuable bench top space
  • Effortless cleaning
  • Display of process status
The Sartorius MA-Series of moisture balances features two infrared metal tube heating elements with a compact footprint and user-friendly design. The BetterClean® design provides for effortless cleaning of all critical instrument parts. The concise touch-screen display panel with easy-to-understand icons guide user through the operation and retrieval of important information. The display panel also features status light that quickly indicates to the user where in the measurement process the sample is. Other features include Mini USB interface, direct data transfer to Microsoft® Windows programs and LED-illuminated sample chamber
The moisture analyzer monitors the drying process and stops the measurement once the sample weight is constant. The built-in weighing system with 1 mg resolution is specifically optimized for use in higher temperature ranges and delivers high measurement accuracy. Typical applications of the MA include the analysis of foods, beverages, pharmaceuticals, chemicals, paper materials and environmental protection products.
772,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Heater Type Readability (g)
Availability Pricing
Sartorius MA37 Infrared Moisture Analyzer, 70G/1mg 1121301 Mfr # MA37-1US
COMPARE
70 Infrared 0.001
Call For Price
Sartorius MA160 Infrared Moisture Analyzer, 200G/1mg 1121303 Mfr # MA160-1US
COMPARE
200 Infrared 0.001
Call For Price
Sartorius MA35 Infrared Moisture Analyzer, 35G/1mg 1121398 Mfr # MA35M-115US
COMPARE
35 Infrared 0.001
₹772,000.00INR / Each