ResinTech VPK Series Cartridge Replacement Kits for High Purity Water System
Reduce operating costs while maintaining peak performance
  • High-capacity cartridges require fewer changes
  • Spin-welded construction with high-purity plastics ensures long service life
  • Available in standard connection and hose nipple configurations
  • Made in the USA!
Use these filtration cartridge replacement kits in your ResinTech, UNIX, Barnstead® PCS® and Vaponics® water purification systems or design a system using wall-mounting brackets and extension tubing (order separately). Permanent joint welds create a hermetic seal and improve longevity in any installation. Nonpigmented ABS plastic housing is durable and produces high-purity laboratory-grade water.
Choose between the standard ARIES connection type and the versatile 3/8" hose nipple, both of which are available in a variety of filter
131,000.00 - ₹185,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Compatible with Media
Availability Pricing
ResinTech VPK Series RO/DI Feed Cartridge Kit for CLiR 3000 High Purity Water System 9928491 Mfr # VPK-4010
COMPARE
(3) 1360 grains ARIES, Unix, Barnstead (D0809, D5027, D8816), Vaponics (MR-1, MR-2) RO/DI Feed, High Purity, Mixed-Bed
₹132,000.00INR / Each
ResinTech VPK Series Tap Feed Cartridge Kit for CLiR 3000 High Purity Water System 9928493 Mfr # VPK-3805
COMPARE
1008 grains / (2) 1360 grains ARIES, Unix, Barnstead (D3805), Vaponics Tap Water Feed, Activated Carbon/Anion, High Purity
₹135,000.00INR / Each
ResinTech VPK Series Filtration Cartridge Kit, Equivalent to Barnstead D4801, (1) Pretreatment, (1) High Capacity, (2) High Purity 9928496 Mfr # VPK-4801
COMPARE
490 grains / 2000 gal. / 1590 grains / (2) 1360 grains ARIES, Unix, Barnstead (D4801), Vaponics Activated Carbon/High Purity, High Capacity, High Purity
₹175,000.00INR / Each
ResinTech VPK Series Filtration Cartridge Kit, Pretreatment, Equaivalent to Barnstead D5028, (1) High Capacity, (2) High Purity 9928502 Mfr # VPK-5028
COMPARE
1008 grains / 1590 grains / (2) 1360 grains ARIES, Unix, Barnstead (D5028), Vaponics Activated Carbon/Anion, High Capacity, High Purity
₹175,000.00INR / Each
ResinTech VPK Series Filtration Cartridge Kit, Pretreatment, Equaivalent to Barnstead D5029, (2) High Purity 9928503 Mfr # VPK-5029
COMPARE
1008 grains / (2) 1360 grains ARIES, Unix, Barnstead (D5029), Vaponics Activated Carbon/Anion, High Purity
₹131,000.00INR / Each
ResinTech VPK Series Filtration Cartridge Kit, Organic Removal, Equaivalent to Barnstead D50228, (2) High Purity, (1) H.P. Low TOC 9928505 Mfr # VPK-50228
COMPARE
1008 grains / 1360 grainss / 1360 grains ARIES, Unix, Barnstead (D50228), Vaponics Activated Carbon/Anion, Low TOC, High Purity
₹185,000.00INR / Each