QLA Small-Volume Glass Dissolution Vessels
Excellent technique when tablets contain only small amounts of active ingredients
  • Glass vessels exhibit good thermal conductivity Highly resistant to scratching
For dissolution tests, taking reliable samples is critical. Use small volume vessels when tablets contain very small amounts of active ingredients or for extended release tablets where small amounts are released that may be difficult to detect. Vessels are manufactured using a unique molded process which provides a precise and consistent inside diameter and spherical radius resulting in more reproducible dissolution test results.
37,300.00 - ₹80,100.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Color Compatible With
Availability Pricing
QLA Small-Volume Vessel for Distek, 100 mL; Amber Glass 4130605 Mfr # GLA100-AD
COMPARE
100 Amber Distek
₹52,400.00INR / Each
QLA Small-Volume Vessel for Distek, 100 mL; Clear Glass 4130606 Mfr # GLA100-DK
COMPARE
100 Clear Distek
₹46,100.00INR / Each
QLA Small-Volume Vessel for Erweka, 100 mL; Clear Glass 4130607 Mfr # GLA100-EW
COMPARE
100 Clear Erweka
₹53,700.00INR / Each
QLA Small-Volume Vessel Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 100 mL; Clear Glass 4130609 Mfr # GLA100-VK
COMPARE
100 Clear Agilent/VanKel/Varian
₹37,300.00INR / Each
QLA Small-Volume Vessel Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 200 mL; Amber Glass 4130611 Mfr # GLA200-AV
COMPARE
200 Amber Agilent/VanKel/Varian
₹80,100.00INR / Each
QLA Small-Volume Vessel for Distek, 200 mL; Clear Glass 4130612 Mfr # GLA200-DK
COMPARE
200 Clear Distek
₹46,600.00INR / Each
QLA Small-Volume Vessel Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 200 mL; Clear Glass 4130613 Mfr # GLA200-VK
COMPARE
200 Clear Agilent/VanKel/Varian
₹47,900.00INR / Each
Showing 1- 7 of 7