QLA Small-Volume Vessel Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 100 mL; Clear Glass
Representative image only.
QLA Small-Volume Vessel Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 100 mL; Clear Glass
| Mfr: GLA100-VK| Item: 4130609
Excellent technique when tablets contain only small amounts of active ingredients
₹37,300.00INR / Each
₹37,300.00INR / Each
Product Description
Key Features
  • Glass vessels exhibit good thermal conductivity Highly resistant to scratching
  • For Agilent/ValKel/Varian baths
  • 100 mL capacity
More About this Item
For dissolution tests, taking reliable samples is critical. Use small volume vessels when tablets contain very small amounts of active ingredients or for extended release tablets where small amounts are released that may be difficult to detect. Vessels are manufactured using a unique molded process which provides a precise and consistent inside diameter and spherical radius resulting in more reproducible dissolution test results.
.
Specifications
  • Capacity (mL)100
  • MaterialGlass
  • ColorClear
  • Compatible WithAgilent/VanKel/Varian
  • DescriptionSmall-Volume Vessel Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 100 mL; Clear Glass