QLA Small-Volume Vessel for Distek, 100 mL; Amber Glass

Representative image only.

QLA Small-Volume Vessel for Distek, 100 mL; Amber Glass

| Mfr: GLA100-AD| Item: 4130605

Excellent technique when tablets contain only small amounts of active ingredients

₹52,400.00INR / Each

₹52,400.00INR / Each
Product Description

Key Features

  • Glass vessels exhibit good thermal conductivity Highly resistant to scratching
  • For Distek baths
  • Amber glass is ideal for light-sensitive compounds
  • 100 mL capacity

More About this Item

For dissolution tests, taking reliable samples is critical. Use small volume vessels when tablets contain very small amounts of active ingredients or for extended release tablets where small amounts are released that may be difficult to detect. Vessels are manufactured using a unique molded process which provides a precise and consistent inside diameter and spherical radius resulting in more reproducible dissolution test results.

.

Specifications
  • Capacity (mL)100
  • MaterialGlass
  • ColorAmber
  • Compatible WithDistek
  • DescriptionSmall Volume Vessel for Distek, 100 mL; Amber Glass
Reviews
Q&A