QLA Small-Volume Vessel for Erweka, 100 mL; Clear Glass

Representative image only.

QLA Small-Volume Vessel for Erweka, 100 mL; Clear Glass

| Mfr: GLA100-EW| Item: 4130607

Excellent technique when tablets contain only small amounts of active ingredients

₹53,700.00INR / Each

₹53,700.00INR / Each
Product Description

Key Features

  • Glass vessels exhibit good thermal conductivity Highly resistant to scratching
  • For Erweka baths
  • 100 mL capacity

More About this Item

For dissolution tests, taking reliable samples is critical. Use small volume vessels when tablets contain very small amounts of active ingredients or for extended release tablets where small amounts are released that may be difficult to detect. Vessels are manufactured using a unique molded process which provides a precise and consistent inside diameter and spherical radius resulting in more reproducible dissolution test results.

.

Specifications
  • Capacity (mL)100
  • MaterialGlass
  • ColorClear
  • Compatible WithErweka
  • DescriptionSmall Volume Vessel for Erweka, 100 mL; Clear Glass
Reviews
Q&A