QLA Distek-Compatible Standard Dissolution Vessels
QLA vessels provide more precise and consistent dissolution results
  • Glass vessels offer good thermal conductivity Highly resistance to scratching
A unique molding process provides a precise and consistent inside diameter and spherical radius resulting in more reproducible dissolution test results.
51,200.00 - ₹117,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Color Material
Availability Pricing
QLA Standard Vessel for Distek, 2000 mL, Amber Glass; Serialized 4130601 Mfr # GLA02K-AD
COMPARE
2000 Amber Glass
₹117,000.00INR / Each
QLA Standard Vessel for Distek, 2000 mL, Clear Glass; Serialized 4130602 Mfr # GLA02K-DK
COMPARE
2000 Clear Glass
₹88,500.00INR / Each
QLA Standard Vessel for Distek, 500 mL, Clear Glass 4130617 Mfr # GLA500-DK
COMPARE
500 Clear Glass
₹56,100.00INR / Each
QLA Standard Vessel for Distek, 1000 mL, Amber Glass; Serialized 4130623 Mfr # GLA900-AD
COMPARE
1000 Amber Glass
₹74,100.00INR / Each
QLA Standard Vessel for Distek, 1000 mL, Clear Glass; Serialized 4130633 Mfr # GLA900-DK
COMPARE
1000 Clear Glass
₹51,200.00INR / Each
QLA Standard Vessel for Distek, 1000 mL, Amber Premier Glass; Serialized 4130634 Mfr # GLA900-DKAP
COMPARE
1000 Amber Premier glass
₹95,100.00INR / Each
QLA Standard Vessel for Distek, 1000 mL, Clear Premier Glass; Serialized 4130635 Mfr # GLA900-DKP
COMPARE
1000 Clear Premier glass
₹69,000.00INR / Each
Showing 1- 7 of 7