QLA Standard Vessel for Distek, 1000 mL, Clear Premier Glass; Serialized

Representative image only.

QLA Standard Vessel for Distek, 1000 mL, Clear Premier Glass; Serialized

| Mfr: GLA900-DKP| Item: 4130635

QLA vessels provide more precise and consistent dissolution results

₹69,000.00INR / Each

₹69,000.00INR / Each
Product Description

Key Features

  • Glass vessels offer good thermal conductivity Highly resistance to scratching
  • Premier glass exhibit tight dimensional tolerances
  • Clear glass
  • 1000 mL capacity
  • Serialized

More About this Item

A unique molding process provides a precise and consistent inside diameter and spherical radius resulting in more reproducible dissolution test results.

Premier glass vessels are manufactured to a tighter tolerence than other vessels to meet the physical dimensions of the UPS. Unique serial number is acid etched on the vessel.

.

Not available for sale in the United States.

Specifications
  • Capacity (mL)1000
  • MaterialPremier glass
  • ColorClear
  • DescriptionStandard Vessel for Distek, 1000 mL, Clear Premier Glass; Serialized
Reviews
Q&A