QLA Standard Vessel for Distek, 2000 mL, Amber Glass; Serialized

Representative image only.

QLA Standard Vessel for Distek, 2000 mL, Amber Glass; Serialized

| Mfr: GLA02K-AD| Item: 4130601

QLA vessels provide more precise and consistent dissolution results

₹117,000.00INR / Each

₹117,000.00INR / Each
Product Description

Key Features

  • Glass vessels offer good thermal conductivity Highly resistance to scratching
  • Amber glass for light-sensitive compounds
  • 2000 mL capacity
  • Serialized

More About this Item

A unique molding process provides a precise and consistent inside diameter and spherical radius resulting in more reproducible dissolution test results.

Standard vessels are manufactured to meet USP physical dimensions. Unique serial number is acid etched on the vessel.

.

Not available for sale in the United States.

Specifications
  • Capacity (mL)2000
  • MaterialGlass
  • ColorAmber
  • DescriptionStandard Vessel for Distek, 2000 mL, Amber Glass; Serialized
Reviews
Q&A