QLA Standard Vessel for Distek, 500 mL, Clear Glass

Representative image only.

QLA Standard Vessel for Distek, 500 mL, Clear Glass

| Mfr: GLA500-DK| Item: 4130617

QLA vessels provide more precise and consistent dissolution results

₹56,100.00INR / Each

₹56,100.00INR / Each
Product Description

Key Features

  • Glass vessels offer good thermal conductivity Highly resistance to scratching
  • Clear glass
  • 500 mL capacity

More About this Item

A unique molding process provides a precise and consistent inside diameter and spherical radius resulting in more reproducible dissolution test results.

Standard vessels are manufactured to meet USP physical dimensions.

.

Not available for sale in the United States.

Specifications
  • Capacity (mL)500
  • MaterialGlass
  • ColorClear
  • DescriptionStandard Vessel for Distek, 500 mL, Clear Glass
Reviews
Q&A