QLA Baskets Shafts for 24.5 mm ID Baskets
QLA Shafts used for Dabigatran Etexilate Mesylate
  • Made of 316 electropolished stainless steel All shafts are laser marked with unique serial numbers
All shafts are crafted from 316 electropolished stainless steel for strength, smoothness and durability. Shafts are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
98,700.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Length (InchShort) Material Compatible With
Availability Pricing
QLA Basket Shaft for Distek 2100 Baths, for 24.5 mm Basket, 16.5"L, 316 SS; Serialized 4130860 Mfr # CUSTSHT-DK
COMPARE
16 1/2 316 stainless steel Distek 2100 baths
₹98,700.00INR / Each
QLA Basket Shaft for Erweka Baths, for 24.5 mm Basket, 20"L, 316 SS; Serialized 4130861 Mfr # CUSTSHT-EW20
COMPARE
20 316 stainless steel Erweka baths
₹98,700.00INR / Each
QLA Basket Shaft for Hanson SR8-Plus Baths, for 24.5 mm Basket, 16"L, 316 SS, 16"; Serialized 4130862 Mfr # CUSTSHT-HR
COMPARE
16 316 stainless steel Hanson SR8-Plus baths
₹98,700.00INR / Each
QLA Basket Shaft for Hanson Vision Baths, for 24.5 mm Basket, 316 SS; Serialized 4130863 Mfr # CUSTSHT-HRV
COMPARE
316 stainless steel Hanson Vision baths
₹98,700.00INR / Each
QLA Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus V-Series Baths, for 24.5 mm Basket, 14.5"L, 316 SS; Serialized 4130864 Mfr # CUSTSHT-VK13
COMPARE
14 1/2 316 stainless steel Agilent/VanKel/Varian V-series baths
₹98,700.00INR / Each
QLA Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus Baths, for 24.5 mm Basket, 15"L, 316 SS; Serialized 4130865 Mfr # CUSTSHT-VK15
COMPARE
15 316 stainless steel Agilent/VanKel/Varian baths
₹98,700.00INR / Each
QLA Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus Baths, for 24.5 mm Basket, 19"L, 316 SS; Serialized 4130866 Mfr # CUSTSHT-VK19
COMPARE
19 316 stainless steel Agilent/VanKel/Varian baths
₹98,700.00INR / Each
Showing 1- 7 of 7