QLA Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus Baths, for 24.5 mm Basket, 19"L, 316 SS; Serialized
Representative image only.
QLA Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus Baths, for 24.5 mm Basket, 19"L, 316 SS; Serialized
| Mfr: CUSTSHT-VK19| Item: 4130866
QLA Shafts used for Dabigatran Etexilate Mesylate
₹98,700.00INR / Each
₹98,700.00INR / Each
Product Description
Key Features
  • Made of 316 electropolished stainless steel All shafts are laser marked with unique serial numbers
  • For Agilent/VanKel/Varian baths
  • 19" long
More About this Item
All shafts are crafted from 316 electropolished stainless steel for strength, smoothness and durability. Shafts are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
.
Specifications
  • Material316 stainless steel
  • Compatible WithAgilent/VanKel/Varian baths
  • Length (InchShort)19
  • Length (cm)48
  • DescriptionBasket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus Baths, for 24.5 mm Basket, 19"L, 316 SS; Serialized