QLA Basket Shaft for Distek 2100 Baths, for 24.5 mm Basket, 16.5"L, 316 SS; Serialized
Representative image only.
QLA Basket Shaft for Distek 2100 Baths, for 24.5 mm Basket, 16.5"L, 316 SS; Serialized
| Mfr: CUSTSHT-DK| Item: 4130860
QLA Shafts used for Dabigatran Etexilate Mesylate
₹98,700.00INR / Each
₹98,700.00INR / Each
Product Description
Key Features
  • Made of 316 electropolished stainless steel All shafts are laser marked with unique serial numbers
  • For Distek 2100 baths
  • 16" long
More About this Item
All shafts are crafted from 316 electropolished stainless steel for strength, smoothness and durability. Shafts are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
.
Specifications
  • Material316 stainless steel
  • Compatible WithDistek 2100 baths
  • Length (InchShort)16 1/2
  • Length (cm)42
  • DescriptionBasket Shaft for Distek 2100 Baths, for 24.5 mm Basket, 16.5"L, 316 SS; Serialized