PYREX® Brand 9826 Culture Tubes
Deep-form threaded caps facilitate handling and sealing
  • Screw cap with PTFE liner
Similar to 9825 culture tubes,except that phenolic caps have a PTFE interface firmly bonded to the rubber liner, providing a highly inert sealing face.
43,900.00 - ₹83,300.00INR / Pkg of 144
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap Style Tube Capacity (mL) Tube Material
Availability Pricing
Pyrex 9826-13 Culture Tube; 9 mL, pack of 144 WW-34572-01 Mfr # 9826-13
COMPARE
Screw Cap 9 Glass
₹65,200.00INR / Pkg of 144
Pyrex 9826-16 Culture Tube; 11 mL, pack of 144 WW-34572-02 Mfr # 9826-16
COMPARE
Screw Cap 11 Glass
₹72,000.00INR / Pkg of 144
Pyrex 9826-16X Culture Tube; 15 mL, pack of 144 WW-34572-03 Mfr # 9826-16X
COMPARE
Screw Cap 15 Glass
₹79,200.00INR / Pkg of 144
Pyrex 9826-16XX Culture Tube; 20 mL, pack of 144 WW-34572-04 Mfr # 9826-16XX
COMPARE
Screw Cap 20 Glass
₹83,300.00INR / Pkg of 144
Pyrex 9826-20 Culture Tube; 25 mL, pack of 96 WW-34572-05 Mfr # 9826-20
COMPARE
Screw Cap 25 Glass
₹69,400.00INR / Pkg of 96
Pyrex 9826-20X Culture Tube; 34 mL, pack of 96 WW-34572-06 Mfr # 9826-20X
COMPARE
Screw Cap 34 Glass
₹72,700.00INR / Pkg of 96
Pyrex 9826-25 Culture Tube; 55 mL, pack of 48 WW-34572-07 Mfr # 9826-25
COMPARE
Screw Cap 55 Glass
₹43,900.00INR / Pkg of 48
Pyrex 9826-25X Culture Tube; 70 mL, pack of 48 WW-34572-08 Mfr # 9826-25X
COMPARE
Screw Cap 70 Glass
₹46,300.00INR / Pkg of 48
Pyrex 9826-16 Culture Tube; 11 mL, case of 288 WW-34572-22 Mfr # 9826-16
COMPARE
Screw Cap 11 Glass
Call For Price
Pyrex 9826-16X Culture Tube; 15 mL, case of 288 WW-34572-23 Mfr # 9826-16X
COMPARE
Screw Cap 15 Glass
Call For Price
Pyrex 9826-20 Culture Tube; 25 mL, case of 192 WW-34572-25 Mfr # 9826-20
COMPARE
Screw Cap 25 Glass
Call For Price
Pyrex 9826-25 Culture Tube; 55 mL, case of 144 WW-34572-27 Mfr # 9826-25
COMPARE
Screw Cap 55 Glass
Call For Price
Showing 1- 12 of 12