Pyrex Brand 9820 Culture Tubes
Rimless
These tubes are rimless for convenient plugging and rack storage. Walls and bottoms are of uniform thickness. Ends are fire polished. The 6-mm, 22-mm OD tubes do not have marking spots.
8,300.00 - ₹80,000.00INR / Pkg of 72
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Outside Diameter (OD) (in) Overall Height (in)
Availability Pricing
Pyrex 9820-10 Culture Tube; 3 mL, pack of 72 WW-34570-01 Mfr # 9820-10
COMPARE
3 25/64 2 61/64
₹8,800.00INR / Pkg of 72
Pyrex 9820-12 Culture Tube; 5 mL, pack of 72 WW-34570-02 Mfr # 9820-12
COMPARE
5 15/32 2 61/64
₹8,800.00INR / Pkg of 72
Pyrex 9820-13 Culture Tube; 9 mL, pack of 72 WW-34570-03 Mfr # 9820-13
COMPARE
9 33/64 3 15/16
₹9,100.00INR / Pkg of 72
Pyrex 9820-16 Culture Tube; 11 mL, pack of 72 WW-34570-04 Mfr # 9820-16
COMPARE
11 5/8 3 15/16
₹11,000.00INR / Pkg of 72
Pyrex 9820-16X Culture Tube; 15 mL, pack of 72 WW-34570-05 Mfr # 9820-16X
COMPARE
15 5/8 4 59/64
₹13,300.00INR / Pkg of 72
Pyrex 9820-16XX Culture Tube; 20 mL, pack of 72 WW-34570-06 Mfr # 9820-16XX
COMPARE
20 5/8 5 29/32
₹14,900.00INR / Pkg of 72
Pyrex 9820-18 Culture Tube; 27 mL, pack of 72 WW-34570-07 Mfr # 9820-18
COMPARE
27 45/64 5 29/32
₹14,200.00INR / Pkg of 72
Pyrex 9820-22 Culture Tube; 50 mL, pack of 72 WW-34570-09 Mfr # 9820-22
COMPARE
50 55/64 6 57/64
₹26,200.00INR / Pkg of 72
Pyrex 9820-25 Culture Tube; 55 mL, pack of 72 WW-34570-10 Mfr # 9820-25
COMPARE
55 63/64 5 29/32
₹27,400.00INR / Pkg of 72
Pyrex 9820-25X Culture Tube; 70 mL, pack of 48 WW-34570-11 Mfr # 9820-25X
COMPARE
70 63/64 7 7/8
₹24,100.00INR / Pkg of 48
Pyrex 9820-6 Culture Tube; 0.5 mL, pack of 72 WW-34570-12 Mfr # 9820-6
COMPARE
0.5 15/64 1 31/32
₹8,300.00INR / Pkg of 72
Pyrex 9820-6 Culture Tube; 0.5 mL, case of 720 WW-34570-32 Mfr # 9820-6
COMPARE
0.5 15/64 1 31/32
₹80,000.00INR / Case of 720
Showing 1- 12 of 12