Motic® Microscope EPI-Fluorescence Accessories
Easily upgrade to an EPI-Fluorescence microscope with interchangeable filters
  • Interchangeable filter cubes cover all routine applications from UV up to NIR excitation
  • Compatible with Motic® AE31 ELITE and BA410/310 microscopes
  • Improved light management offers a powerful excitation for brighter signals
Motic® microscope EPI-fluorescence accessories allow for an easy upgrade to an EPI-Fluorescence microscope by using the established fluorescence attachment and filter sets. Complete system includes microscope, EPI-Fluorescence attachment for corresponding microscope, and fluorescence filter cube filter set. All components sold separately. Choose EPI-Fluorescence attachment based on the type of microscope being used and filter cubes are available with different filter sets for a wide range of applications. Filters are interchangeable between the EPI-fluorescence attachments.
Included in the attachments, the 100-Watt HBO lamphouse with integrated mirror improves light management and offers a powerful excitation for brighter signals. In order to avoid unnecessary fading, the excitation intensity can be adjusted by the power supply.
315,000.00 - ₹1,617,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Compatible With Description
Availability Pricing
Motic Instruments Complete EPI-Fluorescence Attachment for Motic AE31 ELITE Microscope 4892575 Mfr # 1101000202651
COMPARE
Motic AE31 ELITE Microscope Complete EPI-Fluorescence Attachment for Motic AE31 ELITE Microscope
₹1,521,000.00INR / Each
Motic Instruments Complete EPI-Fluorescence Attachment for Motic BA410/310 Microscope 4892576 Mfr # 1101000202171
COMPARE
Motic BA410/310 Microscope Complete EPI-Fluorescence Attachment for Motic BA410/310 Microscope
₹1,617,000.00INR / Each
Motic Instruments Fluorescence Filter Cube for Motic BA410E/AE31E Microscope, DAPI/Hoechst/AlexaFluor 350 4892577 Mfr # 1101000203271
COMPARE
Motic AE31 ELITE and BA410 Microscopes Fluorescence Filter Cube for Motic BA410E/AE31E Microscope, DAPI/Hoechst/AlexaFluor 350
₹315,000.00INR / Each
Motic Instruments Fluorescence Filter Cube for Motic BA410E/AE31E Microscope, EGFP/FITC/Cy2/AlexaFluor 488 4892578 Mfr # 1101000203281
COMPARE
Motic AE31 ELITE and BA410 Microscopes Fluorescence Filter Cube for Motic BA410E/AE31E Microscope, EGFP/FITC/Cy2/AlexaFluor 488
₹315,000.00INR / Each
Motic Instruments Fluorescence Filter cube for Motic BA410E/AE31E Microscope, TRITC/Cy3/TagRFP/AlexaFluor 546 4892579 Mfr # 1101000203291
COMPARE
Motic AE31 ELITE and BA410 Microscopes Fluorescence Filter cube for Motic BA410E/AE31E Microscope, TRITC/Cy3/TagRFP/AlexaFluor 546
₹315,000.00INR / Each