Motic Instruments Complete EPI-Fluorescence Attachment for Motic AE31 ELITE Microscope
Representative image only.
Motic Instruments Complete EPI-Fluorescence Attachment for Motic AE31 ELITE Microscope
Motic Instruments | Mfr: 1101000202651| Item: 4892575
Easily upgrade to an EPI-Fluorescence microscope with interchangeable filters
₹1,521,000.00INR / Each
₹1,521,000.00INR / Each
Buy It Together
Current Item
Motic Instruments Complete EPI-Fluorescence Attachment for Motic AE31 ELITE Microscope
Motic Instruments Complete EPI-Fluorescence Attachment for Motic AE31 ELITE Microscope
4892575
₹1,521,000.00INR / Each
+ =
Total Price 1836000.00 ADD 2 ITEMS TO CART
Product Description
Includes
EPI-Fluorescence attachment with filter cassette, mercury lamp 100W, lamphouse HBO 100W, collector lens and mercury lamp socket for HBO 100W, starter unit HBO 100W, lamp centering tool, and power cord.
Key Features
  • Interchangeable filter cubes cover all routine applications from UV up to NIR excitation
  • Compatible with Motic® AE31 ELITE and BA410/310 microscopes
  • Improved light management offers a powerful excitation for brighter signals
  • Compatible with Motic® AE31 ELITE Microscope
  • Filter sets and microscope sold separately
More About this Item
Motic® microscope EPI-fluorescence accessories allow for an easy upgrade to an EPI-Fluorescence microscope by using the established fluorescence attachment and filter sets. Complete system includes microscope, EPI-Fluorescence attachment for corresponding microscope, and fluorescence filter cube filter set. All components sold separately. Choose EPI-Fluorescence attachment based on the type of microscope being used and filter cubes are available with different filter sets for a wide range of applications. Filters are interchangeable between the EPI-fluorescence attachments.
Included in the attachments, the 100-Watt HBO lamphouse with integrated mirror improves light management and offers a powerful excitation for brighter signals. In order to avoid unnecessary fading, the excitation intensity can be adjusted by the power supply.
Specifications
  • Compatible WithMotic AE31 ELITE Microscope
  • DescriptionComplete EPI-Fluorescence Attachment for Motic AE31 ELITE Microscope