Motic Instruments Fluorescence Filter Cube for Motic BA410E/AE31E Microscope, DAPI/Hoechst/AlexaFluor 350
Representative image only.
Motic Instruments Fluorescence Filter Cube for Motic BA410E/AE31E Microscope, DAPI/Hoechst/AlexaFluor 350
Motic Instruments | Mfr: 1101000203271| Item: 4892577
Easily upgrade to an EPI-Fluorescence microscope with interchangeable filters
₹315,000.00INR / Each
₹315,000.00INR / Each
Buy It Together
Current Item
Motic Instruments Fluorescence Filter Cube for Motic BA410E/AE31E Microscope, DAPI/Hoechst/AlexaFluor 350
Motic Instruments Fluorescence Filter Cube for Motic BA410E/AE31E Microscope, DAPI/Hoechst/AlexaFluor 350
4892577
₹315,000.00INR / Each
+ =
Total Price 1836000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Interchangeable filter cubes cover all routine applications from UV up to NIR excitation
  • Compatible with Motic® AE31 ELITE and BA410/310 microscopes
  • Improved light management offers a powerful excitation for brighter signals
  • Filter sets: DAPI/Hoechst/AlexaFluor 350
  • EPI-Fluorescence attachment and microscope sold separately
More About this Item
Motic® microscope EPI-fluorescence accessories allow for an easy upgrade to an EPI-Fluorescence microscope by using the established fluorescence attachment and filter sets. Complete system includes microscope, EPI-Fluorescence attachment for corresponding microscope, and fluorescence filter cube filter set. All components sold separately. Choose EPI-Fluorescence attachment based on the type of microscope being used and filter cubes are available with different filter sets for a wide range of applications. Filters are interchangeable between the EPI-fluorescence attachments.
Included in the attachments, the 100-Watt HBO lamphouse with integrated mirror improves light management and offers a powerful excitation for brighter signals. In order to avoid unnecessary fading, the excitation intensity can be adjusted by the power supply.
Specifications
  • Compatible WithMotic AE31 ELITE and BA410 Microscopes
  • DescriptionFluorescence Filter Cube for Motic BA410E/AE31E Microscope, DAPI/Hoechst/AlexaFluor 350