Motic Instruments Complete EPI-Fluorescence Attachment for Motic BA410/310 Microscope
Representative image only.
Motic Instruments Complete EPI-Fluorescence Attachment for Motic BA410/310 Microscope
Motic Instruments | Mfr: 1101000202171| Item: 4892576
Easily upgrade to an EPI-Fluorescence microscope with interchangeable filters
₹1,617,000.00INR / Each
₹1,617,000.00INR / Each
Buy It Together
Current Item
Motic Instruments Complete EPI-Fluorescence Attachment for Motic BA410/310 Microscope
Motic Instruments Complete EPI-Fluorescence Attachment for Motic BA410/310 Microscope
4892576
₹1,617,000.00INR / Each
+ =
Total Price 1932000.00 ADD 2 ITEMS TO CART
Product Description
Includes
EPI-Fluorescence attachment with filter cassette, mercury lamp 100W, lamphouse HBO 100W, collector lens and mercury lamp socket, lamp cenerting tool, and power cord.
Key Features
  • Interchangeable filter cubes cover all routine applications from UV up to NIR excitation
  • Compatible with Motic® AE31 ELITE and BA410/310 microscopes
  • Improved light management offers a powerful excitation for brighter signals
  • Compatible with Motic® BA410/310 Microscope
  • Filter sets and microscope sold separately
More About this Item
Motic® microscope EPI-fluorescence accessories allow for an easy upgrade to an EPI-Fluorescence microscope by using the established fluorescence attachment and filter sets. Complete system includes microscope, EPI-Fluorescence attachment for corresponding microscope, and fluorescence filter cube filter set. All components sold separately. Choose EPI-Fluorescence attachment based on the type of microscope being used and filter cubes are available with different filter sets for a wide range of applications. Filters are interchangeable between the EPI-fluorescence attachments.
Included in the attachments, the 100-Watt HBO lamphouse with integrated mirror improves light management and offers a powerful excitation for brighter signals. In order to avoid unnecessary fading, the excitation intensity can be adjusted by the power supply.
Specifications
  • Compatible WithMotic BA410/310 Microscope
  • DescriptionComplete EPI-Fluorescence Attachment for Motic BA410/310 Microscope