KIMAX Narrow Mouth Reservoir Bottles, Glass
Easily discharge liquids via a bottom glass hose outlet
  • Easy to read white enamel marking spot
  • Conforms to ASTM E438, Type I, Class A
Bottom glass hose connection outlet is fused to the bottle allows you to easily to store and discharge liquids. The spigot on the 250- and 500-mL bottles accept 5/16" ID flexible tubing; large size bottles accept 3/8" ID tubing. Bottles have permanent white enamel marking spot.
48,700.00 - ₹432,000.00INR / Case of 6
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Cap / Stopper Size Material
Availability Pricing
DWK Life Sciences (Kimble) 14607-250 Reservoir with Bottom Hose Outlet, Narrow-Mouth for Stopper; 250 mL 3454101 Mfr # 14607-250
COMPARE
0.25 #2 Glass
₹172,000.00INR / Case of 6
DWK Life Sciences (Kimble) 14607-500 Reservoir with Bottom Hose Outlet, Narrow-Mouth for Stopper, 500 ML 3454103 Mfr # 14607-500
COMPARE
0.5 #4 Glass
₹49,000.00INR / Each
DWK Life Sciences (Kimble) 14607-1000 Reservoir with Bottom Hose Outlet, Narrow-Mouth for Stopper, 1L 3454105 Mfr # 14607-1000
COMPARE
1 #6 Glass
₹48,700.00INR / Each
DWK Life Sciences (Kimble) 14607-2000 Reservoir with Bottom Hose Outlet, Narrow-Mouth for Stopper, 2L 3454107 Mfr # 14607-2000
COMPARE
2 #6 Glass
₹71,600.00INR / Each
DWK Life Sciences (Kimble) 14607-10000 Reservoir with Bottom Hose Outlet, Narrow-Mouth for Stopper, 10L 3454111 Mfr # 14607-10000
COMPARE
10 #11.5 Glass
₹209,000.00INR / Each
DWK Life Sciences (Kimble) 14607-20000 Reservoir with Bottom Hose Outlet, Narrow-Mouth for Stopper, 20L 3454113 Mfr # 14607-20000
COMPARE
20 #11.5 Glass
₹432,000.00INR / Each