Keck Plastic Clips
Our most economical standard taper clips
  • Securely holds conical joints under pressure
  • Safe to at 302 °>F (150 °>C)
These economical standard taper clips securely hold conical joints under pressure without damaging glass. The polyoxymethylene (POM) acetal resin clips are safe to 302°>F (150°>C). Resistant to concentrated alkalies and dilute acids (not resistant to halogen gases and concentrated acids).
9,600.00 - ₹28,500.00INR / Pkg of 12
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Clamp Material Clamp Type
Availability Pricing
Keck Standard Taper Clips, Joint Size 10, Green, Pk/12 0683990 Mfr # MAKE-UP
COMPARE
Polyoxymethylene (POM) Acetal Standard Taper, Joint Size 10
₹10,500.00INR / Pkg of 12
Keck Standard Taper Clips, Joint Size 12, Violet, Pk/12 0683995 Mfr # MAKE-UP
COMPARE
Polyoxymethylene (POM) Acetal Standard Taper, Joint Size 12
₹10,800.00INR / Pkg of 12
Keck Standard Taper Clips Assortment, 24/Pk 0684000 Mfr # MAKE-UP
COMPARE
Polyoxymethylene (POM) Acetal Standard Taper, Joint Sizes: 14, 19, 24 and 29
₹14,500.00INR / Pkg of 24
Keck Standard Taper Clips, Joint Size 14, Yellow, Pk/12 0684005 Mfr # MAKE-UP
COMPARE
Polyoxymethylene (POM) Acetal Standard Taper, Joint Size 14
₹11,600.00INR / Pkg of 12
Keck Standard Taper Clips, Joint Size 19, Blue, Pk/12 0684010 Mfr # MAKE-UP
COMPARE
Polyoxymethylene (POM) Acetal Standard Taper, Joint Size 19
₹11,800.00INR / Pkg of 12
Keck Standard Taper Clips, Joint Size 24, Green, Pk/12 0684015 Mfr # MAKE-UP
COMPARE
Polyoxymethylene (POM) Acetal Standard Taper, Joint Size 24
₹17,900.00INR / Pkg of 12
Keck Standard Taper Clips, Joint Size 29, Red, Pk/12 0684020 Mfr # MAKE-UP
COMPARE
Polyoxymethylene (POM) Acetal Standard Taper, Joint Size 29
₹17,900.00INR / Pkg of 12
Keck Standard Taper Clips, Joint Size 34, ORange , Pk/12 0684025 Mfr # MAKE-UP
COMPARE
Polyoxymethylene (POM) Acetal Standard Taper, Joint Size 34
₹12,700.00INR / Pkg of 12
Keck Standard Taper Clips, Joint Size 40, Yellow, Pk/12 0684030 Mfr # MAKE-UP
COMPARE
Polyoxymethylene (POM) Acetal Standard Taper, Joint Size 40
₹14,400.00INR / Pkg of 12
Keck Standard Taper Clips, Joint Size 45, Brown, Pk/12 0684035 Mfr # MAKE-UP
COMPARE
Polyoxymethylene (POM) Acetal Standard Taper, Joint Size 45
₹9,600.00INR / Pkg of 12
Keck Standard Taper Clip Assortment, Pk/27 0684050 Mfr # MAKE-UP
COMPARE
Polyoxymethylene (POM) Acetal Standard Taper, Joint Sizes: 10, 12, 14, 19, 24, 29, 34, 40, 45
₹28,500.00INR / Pkg of 27
Showing 1- 11 of 11