Keck Standard Taper Clip Assortment, Pk/27
Representative image only.
Keck Standard Taper Clip Assortment, Pk/27
Keck | Mfr: MAKE-UP| Item: 0684050
Our most economical standard taper clips
₹28,500.00INR / Pkg of 27
₹28,500.00INR / Pkg of 27
Product Description
Key Features
  • Securely holds conical joints under pressure
  • Safe to at 302 °>F (150 °>C)
  • Contains 3 each of standard taper joint clips in the following sizes: 10, 12, 14, 19, 24, 29, 34, 40, 45
More About this Item
These economical standard taper clips securely hold conical joints under pressure without damaging glass. The polyoxymethylene (POM) acetal resin clips are safe to 302°>F (150°>C). Resistant to concentrated alkalies and dilute acids (not resistant to halogen gases and concentrated acids).
Specifications
  • Clamp MaterialPolyoxymethylene (POM) Acetal
  • Clamp TypeStandard Taper, Joint Sizes: 10, 12, 14, 19, 24, 29, 34, 40, 45
  • ColorAssorted
  • DescriptionStandard Taper Clips Assortment, Pk/27