Keck Standard Taper Clips, Joint Size 40, Yellow, Pk/12
Representative image only.
Keck Standard Taper Clips, Joint Size 40, Yellow, Pk/12
Keck | Mfr: MAKE-UP| Item: 0684030
Our most economical standard taper clips
₹14,400.00INR / Pkg of 12
₹14,400.00INR / Pkg of 12
Product Description
Key Features
  • Securely holds conical joints under pressure
  • Safe to at 302 °>F (150 °>C)
More About this Item
These economical standard taper clips securely hold conical joints under pressure without damaging glass. The polyoxymethylene (POM) acetal resin clips are safe to 302°>F (150°>C). Resistant to concentrated alkalies and dilute acids (not resistant to halogen gases and concentrated acids).
Specifications
  • Clamp MaterialPolyoxymethylene (POM) Acetal
  • Clamp TypeStandard Taper, Joint Size 40
  • ColorYellow
  • DescriptionStandard Taper Clips, Joint Size 40, Yellow, Pk/12