Keck Standard Taper Clips Assortment, 24/Pk
Representative image only.
Keck Standard Taper Clips Assortment, 24/Pk
Keck | Mfr: MAKE-UP| Item: 0684000
Our most economical standard taper clips
₹14,500.00INR / Pkg of 24
₹14,500.00INR / Pkg of 24
Product Description
Key Features
  • Securely holds conical joints under pressure
  • Safe to at 302 °>F (150 °>C)
  • Contains 6 each of standard taper joint clips in the following sizes: 14, 19, 24, 29
More About this Item
These economical standard taper clips securely hold conical joints under pressure without damaging glass. The polyoxymethylene (POM) acetal resin clips are safe to 302°>F (150°>C). Resistant to concentrated alkalies and dilute acids (not resistant to halogen gases and concentrated acids).
Specifications
  • Clamp MaterialPolyoxymethylene (POM) Acetal
  • Clamp TypeStandard Taper, Joint Sizes: 14, 19, 24 and 29
  • ColorAssorted
  • DescriptionStandard Taper Clips Assortment, 24/Pk