IKA MultiDrive Basic Grinding Mill Accessories

Choose from a variety of vessels and other accessories to expand the capabilities of your IKA® grinding mill
  • Compatible with IKA grinder mill 28711-53
Choose from a variety of cutters and beaters to expand the capabilities of your IKA MultiDrive grinding mill
63,500.00 - ₹166,000.00INR / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Material Sample Capacity (mL) Description
Availability Pricing
IKA 0025003961 Grinding Mill Vessel with Mixing Knife, 2000 mL 2871154 Mfr # 0025003961
COMPARE
Stainless steel 2000 Grinding Mill Vessel with Mixing Knife, 2000 mL
₹63,500.00INR / Each
IKA 0020023169 Standard Mixing Knife, Stainless Steel 2871157 Mfr # 0020023169
COMPARE
Stainless steel 2000 Standard Mixing Knife, Stainless Steel
₹65,400.00INR / Each
IKA 0020023167 Star Shaped Beater, Stainless Steel, 2000 mL 2871159 Mfr # 0020023167
COMPARE
Stainless steel 250 Star Shaped Cutter, Stainless Steel
₹76,300.00INR / Each
IKA 0020023168 Wing Style Beater, Tungsten Carbide, 250 mL 2871160 Mfr # 0020023168
COMPARE
Tungsten carbide 250 Beater for Grinding Mill, Tungsten Carbide
₹166,000.00INR / Each
IKA 0020023171 Star Shaped Beater, Stainless Steel, 400 mL 2871162 Mfr # 0020023171
COMPARE
Stainless steel 400 Star Shaped Cutter, Stainless Steel
₹85,300.00INR / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat