IKA 0025003961 Grinding Mill Vessel with Mixing Knife, 2000 mL

Representative image only.

IKA 0025003961 Grinding Mill Vessel with Mixing Knife, 2000 mL

IKA | Mfr: 0025003961| Item: 2871154

Choose from a variety of vessels and other accessories to expand the capabilities of your IKA® grinding mill

₹63,500.00INR / Each

₹63,500.00INR / Each
Product Description

Includes

Stainless steel mixing knife

Key Features

  • Compatible with IKA grinder mill 28711-53
  • Great for wet grinding applications, such as meat, sausage, soy beans, silage, vegetables
  • Dishwasher safe

More About this Item

Choose from a variety of cutters and beaters to expand the capabilities of your IKA MultiDrive grinding mill

Two year warranty
Specifications
  • Sample Capacity (mL)2000
  • MaterialStainless steel
  • DescriptionGrinding Mill Vessel with Mixing Knife, 2000 mL
Reviews
Q&A