IKA 0020023168 Wing Style Beater, Tungsten Carbide, 250 mL
Representative image only.
IKA 0020023168 Wing Style Beater, Tungsten Carbide, 250 mL
IKA | Mfr: 0020023168| Item: 2871160
Choose from a variety of vessels and other accessories to expand the capabilities of your IKA® grinding mill
₹166,000.00INR / Each
₹166,000.00INR / Each
Product Description
Key Features
  • Compatible with IKA grinder mill 28711-53
  • Suitable for harder materials up to Mohs hardness 9
  • Compatible with IKA MultiDrive MI 250 (28711-55)
More About this Item
Choose from a variety of cutters and beaters to expand the capabilities of your IKA MultiDrive grinding mill
Two year warranty
Specifications
  • Sample Capacity (mL)250
  • MaterialTungsten carbide
  • DescriptionBeater for Grinding Mill, Tungsten Carbide