IKA 0020023171 Star Shaped Beater, Stainless Steel, 400 mL
Representative image only.
IKA 0020023171 Star Shaped Beater, Stainless Steel, 400 mL
IKA | Mfr: 0020023171| Item: 2871162
Choose from a variety of vessels and other accessories to expand the capabilities of your IKA® grinding mill
₹85,300.00INR / Each
₹85,300.00INR / Each
Product Description
Key Features
  • Compatible with IKA grinder mill 28711-53
  • Suitable for fibrous materials, such as paper and plant material
  • Compatible with IKA MultiDrive MI 400 (28711-56)
More About this Item
Choose from a variety of cutters and beaters to expand the capabilities of your IKA MultiDrive grinding mill
Two year warranty
Specifications
  • Sample Capacity (mL)400
  • MaterialStainless steel
  • DescriptionStar Shaped Cutter, Stainless Steel