Idex® VHP (Very High Pressure) Chromatography Ferrules
Ferrules for use with VHP fittings
  • Designed to withstand extreme pressures
  • Ideal for UHPLC applications
These fittings ensure reliable connections and performance for your instruments.
3,500.00 - ₹39,800.00INR / Pkg of 10
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Material Compatible With Description
Availability Pricing
Idex VHP LiteTouch® Ferrule, PK, 1/16" OD Tubing, 10-32 Coned; 10/PK 0201465 Mfr # PK-100X
COMPARE
PK Use with VHP PK nut 02014-66 VHP LiteTouch® Ferrule, PK, 1/16" OD Tubing, 10-32 Coned; 10/PK
₹26,000.00INR / Pkg of 10
Idex VHP MicroFerrule, Natural PK, 1/32" OD Tubing, 5/16-24 Coned; 1/EA 0202255 Mfr # PK-112
COMPARE
PK Use with VHP micro fittings 02022-53 and -54 VHP MicroFerrule, Natural PK, 1/32" OD Tubing, 5/16-24 Coned; 1/EA
₹3,500.00INR / Each
Idex VHP LiteTouch® Ferrule, PK, 1/32" OD Tubing, 10-32 Coned; 1/EA 0202259 Mfr # PK-132
COMPARE
PK Use with VHP PK nut 02014-66 VHP LiteTouch® Ferrule, PK, 1/32" OD Tubing, 10-32 Coned; 1/EA
₹4,200.00INR / Each
Idex VHP LiteTouch® Ferrule, PK, 1/32" OD Tubing, 10-32 Coned; 10/PK 0202260 Mfr # PK-132X
COMPARE
PK Use with VHP PK nut 02014-66 VHP LiteTouch® Ferrule, PK, 1/32" OD Tubing, 10-32 Coned; 10/PK
₹39,800.00INR / Pkg of 10
Idex VHP MicroFerrule, Natural PK, 360 µm OD Tubing, 5/16-24 Coned; 1/EA 0202261 Mfr # PK-152
COMPARE
PK Use with VHP micro fittings 02022-53 and -54 VHP MicroFerrule, Natural PK, 360 µm OD Tubing, 5/16-24 Coned; 1/EA
₹3,500.00INR / Each