Idex VHP MicroFerrule, Natural PK, 360 µm OD Tubing, 5/16-24 Coned; 1/EA

Representative image only.

Idex VHP MicroFerrule, Natural PK, 360 µm OD Tubing, 5/16-24 Coned; 1/EA

Idex | Mfr: PK-152| Item: 0202261

Ferrules for use with VHP fittings

₹3,500.00INR / Each

₹3,500.00INR / Each
Product Description

Key Features

  • Designed to withstand extreme pressures
  • Ideal for UHPLC applications
  • Use with VHP micro fittings 02022-53 and -54
  • Max pressure of 15,000 psi (1035 bar)
  • For 360 µm OD tubing
  • 1 year warranty

More About this Item

These fittings ensure reliable connections and performance for your instruments.

1 Year Warranty
Specifications
  • MaterialPK
  • Compatible WithUse with VHP micro fittings 02022-53 and -54
  • DescriptionVHP MicroFerrule, Natural PK, 360 µm OD Tubing, 5/16-24 Coned; 1/EA
  • Warranty1 year
Reviews
Q&A