Idex VHP LiteTouch® Ferrule, PK, 1/32" OD Tubing, 10-32 Coned; 10/PK
Representative image only.
Idex VHP LiteTouch® Ferrule, PK, 1/32" OD Tubing, 10-32 Coned; 10/PK
Idex | Mfr: PK-132X| Item: 0202260
Ferrules for use with VHP fittings
₹39,800.00INR / Pkg of 10
₹39,800.00INR / Pkg of 10
Product Description
Key Features
  • Designed to withstand extreme pressures
  • Ideal for UHPLC applications
  • Use with VHP PK nut 02014-66
  • Max of pressure 16,500 psi (1140 bar)
  • For 1/32" OD tubing
  • 10/PK
  • 1 year warranty
More About this Item
These fittings ensure reliable connections and performance for your instruments.
1 Year Warranty
Specifications
  • MaterialPK
  • Compatible WithUse with VHP PK nut 02014-66
  • DescriptionVHP LiteTouch® Ferrule, PK, 1/32" OD Tubing, 10-32 Coned; 10/PK