Idex VHP MicroFerrule, Natural PK, 1/32" OD Tubing, 5/16-24 Coned; 1/EA
Representative image only.
Idex VHP MicroFerrule, Natural PK, 1/32" OD Tubing, 5/16-24 Coned; 1/EA
Idex | Mfr: PK-112| Item: 0202255
Ferrules for use with VHP fittings
₹3,500.00INR / Each
₹3,500.00INR / Each
Product Description
Key Features
  • Designed to withstand extreme pressures
  • Ideal for UHPLC applications
  • Use with VHP micro fittings 02022-53 and -54
  • Max pressure of 15,000 psi (1035 bar)
  • For 1/32" OD tubing
  • 1 year warranty
More About this Item
These fittings ensure reliable connections and performance for your instruments.
1 Year Warranty
Specifications
  • MaterialPK
  • Compatible WithUse with VHP micro fittings 02022-53 and -54
  • DescriptionVHP MicroFerrule, Natural PK, 1/32" OD Tubing, 5/16-24 Coned; 1/EA