Idex® Microferrules for MicroTight® Female Nuts

Make superior fingertight connections with capillary tubing
  • Constructed of durable PEEK
Select microferrules for 360 µm, 1/32", 1/16" OD tubing or MicroTight tubing sleeves.
2,700.00 - ₹5,600.00INR / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Color Description
Availability Pricing
Idex MicroTight® Ferrule, Natural PEEK, 1/32" OD Tubing, 5/16-24 Coned; 1/EA 0201873 Mfr # F-112
COMPARE
Natural MicroTight® Ferrule, Natural PEEK, 1/32" OD Tubing, 5/16-24 Coned; 1/EA
₹2,800.00INR / Each
Idex MicroTight® Ferrule, Natural PEEK, 1/16" OD Tubing, 5/16-24 Coned; 1/EA 0201892 Mfr # F-132
COMPARE
Natural MicroTight® Ferrule, Natural PEEK, 1/16" OD Tubing, 5/16-24 Coned; 1/EA
₹5,600.00INR / Each
Idex MicroTight® Ferrule, Natural PEEK, 360 µm OD Tubing, 5/16-24 Coned; 1/EA 0201895 Mfr # F-152
COMPARE
Natural MicroTight® Ferrule, Natural PEEK, 360 µm OD Tubing, 5/16-24 Coned; 1/EA
₹2,700.00INR / Each
Idex MicroTight® Ferrule, Black PEEK, Tubing Sleeves, 5/16-24 Coned; 1/EA 0201898 Mfr # F-172
COMPARE
Black MicroTight® Ferrule, Black PEEK, Tubing Sleeves, 5/16-24 Coned; 1/EA
₹2,800.00INR / Each
Idex MicroTight® Ferrule Plug, Black PEEK, 5/16-24 Coned; 1/EA 0201919 Mfr # P-116
COMPARE
Black MicroTight® Ferrule Plug, Black PEEK, 5/16-24 Coned; 1/EA
₹5,600.00INR / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat