Idex MicroTight® Ferrule, Natural PEEK, 1/32" OD Tubing, 5/16-24 Coned; 1/EA

Representative image only.

Idex MicroTight® Ferrule, Natural PEEK, 1/32" OD Tubing, 5/16-24 Coned; 1/EA

Idex | Mfr: F-112| Item: 0201873

Make superior fingertight connections with capillary tubing

₹2,800.00INR / Each

₹2,800.00INR / Each
Product Description

Key Features

  • Constructed of durable PEEK
  • For 1/32" OD tubing
  • 1 year warranty

More About this Item

Select microferrules for 360 µm, 1/32", 1/16" OD tubing or MicroTight tubing sleeves.

1 Year Warranty
Specifications
  • MaterialPEEK
  • ColorNatural
  • Compatible With5/16-24 coned, for 1/32" OD tubing
  • DescriptionMicroTight® Ferrule, Natural PEEK, 1/32" OD Tubing, 5/16-24 Coned; 1/EA
  • Warranty1 year
Reviews
Q&A