Idex MicroTight® Ferrule, Black PEEK, Tubing Sleeves, 5/16-24 Coned; 1/EA
Representative image only.
Idex MicroTight® Ferrule, Black PEEK, Tubing Sleeves, 5/16-24 Coned; 1/EA
Idex | Mfr: F-172| Item: 0201898
Make superior fingertight connections with capillary tubing
₹2,800.00INR / Each
₹2,800.00INR / Each
Product Description
Key Features
  • Constructed of durable PEEK
  • For tubing sleeves
  • 1 year warranty
More About this Item
Select microferrules for 360 µm, 1/32", 1/16" OD tubing or MicroTight tubing sleeves.
1 Year Warranty
Specifications
  • MaterialPEEK
  • ColorBlack
  • Compatible With5/16-24 coned, for tubing sleeves
  • DescriptionMicroTight® Ferrule, Black PEEK, Tubing Sleeves, 5/16-24 Coned; 1/EA